Assay Details
| Gene Symbol: FBXO34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561319.2 | XP_005561376.1 | 2 | 687 | 156 | |
| XM_005561320.2 | XP_005561377.1 | 2 | 622 | 156 | ||
More Information
Set Membership: |
|

FBXO34
Genomic Map