Assay Details
| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555756.2 | XP_005555813.1 | 2 - 3 | 856 | 68 | |
| XM_005555757.2 | XP_005555814.1 | 2 - 3 | 856 | 68 | ||
More Information
Set Membership: |
|

ARSJ
Genomic Map