Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551192.2 | XP_005551249.1 | 2 - 3 | 536 | 68 | |
| XM_015448295.1 | XP_015303781.1 | 2 - 3 | 489 | 68 | ||
More Information
Set Membership: |
|

TMUB1
Genomic Map