Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572201.1 | XP_005572258.1 | 2 - 3 | 325 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005572201.1 transcripts.
Set Membership: |
|

FAM222A
Genomic Map