Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593773.2 | XP_005593830.1 | 2 - 3 | 997 | 117 | |
| XM_005593774.2 | XP_005593831.1 | 2 - 3 | 1022 | 117 | ||
More Information
Set Membership: |
|

ASB12
Genomic Map