Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543331.2 | XP_005543388.1 | 1 - 2 | 665 | 85 | |
| GenBank mRNA | AB125158.1 | - | 1 - 2 | 271 | 85 | |
| AB170802.1 | - | 1 - 2 | 271 | 85 | ||
More Information
Set Membership: |
|

CPT2
Genomic Map