Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594276.1 | XP_005594333.1 | 2 - 3 | 694 | 132 | |
| XM_005594277.2 | XP_005594334.1 | 2 - 3 | 694 | 132 | ||
More Information
Set Membership: |
|

SERPINA7
Genomic Map