Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574315.2 | XP_005574372.1 | 1 - 2 | 1477 | 107 | |
| GenBank mRNA | AB125162.1 | - | 1 - 2 | 597 | 107 | |
| DQ074803.1 | - | 1 - 2 | 262 | 107 | ||
More Information
Set Membership: |
|

CYP27A1
Genomic Map