Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542297.2 | XP_005542354.1 | 2 - 3 | 494 | 138 | |
| XM_005542298.2 | XP_005542355.1 | 1 - 2 | 1782 | 138 | ||
| XM_015430975.1 | XP_015286461.1 | 1 - 2 | 1778 | 138 | ||
| XM_015430980.1 | XP_015286466.1 | 2 - 3 | 1554 | 138 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005542297.2, XM_005542298.2, XM_015430975.1, XM_015430980.1 transcripts.
Set Membership: |
|

TAFA3
Genomic Map