Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542297.2 | XP_005542354.1 | 4 - 5 | 755 | 95 | |
| XM_015430975.1 | XP_015286461.1 | 3 - 4 | 2039 | 95 | ||
| XM_015430980.1 | XP_015286466.1 | 4 - 5 | 1815 | 95 | ||
More Information
Set Membership: |
|

TAFA3
Genomic Map