Assay Details
| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319534.1 | NP_001306463.1 | 1 - 2 | 634 | 112 | |
| GenBank mRNA | AB060227.1 | - | 1 - 2 | 732 | 112 | |
| AB096992.1 | - | 1 - 2 | 634 | 112 | ||
More Information
Set Membership: |
|

NAIF1
Genomic Map