Assay Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586312.2 | XP_005586369.1 | 4 - 5 | 810 | 143 | |
| XM_005586313.2 | XP_005586370.1 | 3 - 4 | 627 | 143 | ||
| XM_005586314.2 | XP_005586371.1 | 4 - 5 | 665 | 143 | ||
More Information
Set Membership: |
|

DCUN1D2
Genomic Map