Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541076.2 | XP_005541133.1 | 6 - 7 | 1622 | 87 | |
| XM_005541077.2 | XP_005541134.1 | 6 - 7 | 1479 | 87 | ||
| XM_015450946.1 | XP_015306432.1 | 5 - 6 | 2894 | 87 | ||
| XM_015450951.1 | XP_015306437.1 | 6 - 7 | 1344 | 87 | ||
More Information
Set Membership: |
|

TRIM17
Genomic Map