Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550282.2 | XP_005550339.1 | 3 - 4 | 808 | 94 | |
| XM_005550283.2 | XP_005550340.1 | 3 - 4 | 1072 | 94 | ||
| XM_005550284.2 | XP_005550341.1 | 3 - 4 | 841 | 94 | ||
| XM_005550285.2 | XP_005550342.1 | 3 - 4 | 808 | 94 | ||
| XM_005550286.2 | XP_005550343.1 | 3 - 4 | 808 | 94 | ||
| XM_015447806.1 | XP_015303292.1 | 2 - 3 | 831 | 94 | ||
| XM_015447807.1 | XP_015303293.1 | 2 - 3 | 887 | 94 | ||
More Information
Set Membership: |
|

STEAP2
Genomic Map