Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592894.2 | XP_005592951.1 | 6 - 7 | 670 | 104 | |
| XM_005592895.2 | XP_005592952.1 | 6 - 7 | 670 | 104 | ||
| XM_005592896.2 | XP_005592953.1 | 5 - 6 | 537 | 104 | ||
| XM_015443596.1 | XP_015299082.1 | 5 - 6 | 588 | 104 | ||
| XM_015443597.1 | XP_015299083.1 | 6 - 7 | 656 | 104 | ||
| XM_015443598.1 | XP_015299084.1 | 5 - 6 | 611 | 104 | ||
More Information
Set Membership: |
|

ARSF
Genomic Map