Assay Details
| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543434.2 | XP_005543491.1 | 7 - 8 | 911 | 108 | |
| XM_005543436.2 | XP_005543493.1 | 7 - 8 | 912 | 108 | ||
| XM_005543437.2 | XP_005543494.1 | 7 - 8 | 912 | 108 | ||
| XM_005543438.2 | XP_005543495.1 | 7 - 8 | 884 | 108 | ||
| XM_015452583.1 | XP_015308069.1 | 7 - 8 | 884 | 108 | ||
| XM_015452587.1 | XP_015308073.1 | 8 - 9 | 990 | 108 | ||
| XM_015452589.1 | XP_015308075.1 | 6 - 7 | 759 | 108 | ||
More Information
Set Membership: |
|

AGBL4
Genomic Map