Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015435400.1 | XP_015290886.1 | 1 - 2 | 948 | 124 | |
| XM_015435401.1 | XP_015290887.1 | 1 - 2 | 948 | 124 | ||
| XM_015435402.1 | XP_015290888.1 | 1 - 2 | 948 | 124 | ||
| XM_015435403.1 | XP_015290889.1 | 1 - 2 | 949 | 124 | ||
| XM_015435404.1 | XP_015290890.1 | 1 - 2 | 948 | 124 | ||
| XM_015435405.1 | XP_015290891.1 | 1 - 2 | 948 | 124 | ||
| XM_015435406.1 | XP_015290892.1 | 1 - 2 | 950 | 124 | ||
| XM_015435407.1 | XP_015290893.1 | 1 - 2 | 949 | 124 | ||
| XM_015435408.1 | XP_015290894.1 | 2 - 3 | 970 | 124 | ||
| XM_015435409.1 | XP_015290895.1 | 1 - 2 | 952 | 124 | ||
| XM_015435410.1 | XP_015290896.1 | 1 - 2 | 948 | 124 | ||
| XM_015435412.1 | XP_015290898.1 | 1 - 2 | 952 | 124 | ||
| XM_015435413.1 | XP_015290899.1 | 1 - 2 | 951 | 124 | ||
| XM_015435415.1 | XP_015290901.1 | 1 - 2 | 954 | 124 | ||
More Information
Set Membership: |
|

ART5
Genomic Map