Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580225.2 | XP_005580282.1 | 5 - 6 | 1241 | 89 | |
| XM_005580227.2 | XP_005580284.1 | 5 - 6 | 1198 | 89 | ||
More Information
Set Membership: |
|

B3GAT1
Genomic Map