Assay Details
| Gene Symbol: NIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558916.2 | XP_005558973.1 | 3 - 4 | 386 | 91 | |
| XM_015452513.1 | XP_015307999.1 | 2 - 3 | 295 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015452513.1 transcripts.
Set Membership: |
|

NIPA1
Genomic Map