Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567460.2 | XP_005567517.1 | 2 - 3 | 357 | 114 | |
| XM_005567461.2 | XP_005567518.1 | 2 - 3 | 306 | 114 | ||
| XM_015457261.1 | XP_015312747.1 | 2 - 3 | 215 | 114 | ||
More Information
Set Membership: |
|

MB
Genomic Map