Assay Details
| Gene Symbol: ZNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594873.2 | XP_005594930.1 | 8 - 9 | 724 | 137 | |
| XM_005594875.2 | XP_005594932.1 | 8 - 9 | 722 | 137 | ||
| XM_005594877.2 | XP_005594934.1 | 8 - 9 | 723 | 137 | ||
| XM_005594878.2 | XP_005594935.1 | 8 - 9 | 723 | 137 | ||
| XM_015444179.1 | XP_015299665.1 | 7 - 8 | 668 | 137 | ||
| XM_015444180.1 | XP_015299666.1 | 8 - 9 | 724 | 137 | ||
| XM_015444181.1 | XP_015299667.1 | 8 - 9 | 722 | 137 | ||
More Information
Set Membership: |
|

ZNF185
Genomic Map