Assay Details
| Gene Symbol: ATXN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283531.1 | NP_001270460.1 | 9 - 10 | 1364 | 83 | |
| GenBank mRNA | AB169762.1 | - | 9 - 10 | 1430 | 83 | |
| AB171571.1 | - | 9 - 10 | 1364 | 83 | ||
More Information
Set Membership: |
|

ATXN10
Genomic Map