Assay Details
| Gene Symbol: MOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285112.1 | NP_001272041.1 | 2 - 3 | 305 | 92 | |
| XM_005562236.2 | XP_005562293.1 | 2 - 3 | 354 | 92 | ||
| XM_015453261.1 | XP_015308747.1 | 2 - 3 | 354 | 92 | ||
| XM_015453263.1 | XP_015308749.1 | 2 - 3 | 354 | 92 | ||
| GenBank mRNA | AB168495.1 | - | 2 - 3 | 173 | 92 | |
| AB169545.1 | - | 2 - 3 | 305 | 92 | ||
More Information
Set Membership: |
|

MOK
Genomic Map