Assay Details
Gene Symbol: SACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585474.2 | XP_005585531.1 | 5 - 6 | 1004 | 95 | |
XM_005585475.2 | XP_005585532.1 | 6 - 7 | 675 | 95 | ||
XM_005585476.2 | XP_005585533.1 | 4 - 5 | 404 | 95 | ||
XM_005585477.2 | XP_005585534.1 | 5 - 6 | 975 | 95 | ||
XM_015439055.1 | XP_015294541.1 | 5 - 6 | 975 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
