Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574693.2 | XP_005574750.1 | 6 - 7 | 3555 | 94 | |
| XM_005574694.2 | XP_005574751.1 | 6 - 7 | 3567 | 94 | ||
| XM_005574696.2 | XP_005574753.1 | 4 - 5 | 3409 | 94 | ||
| XM_005574697.2 | XP_005574754.1 | 4 - 5 | 3347 | 94 | ||
| XM_015432996.1 | XP_015288482.1 | 4 - 5 | 4634 | 94 | ||
| XM_015432997.1 | XP_015288483.1 | 4 - 5 | 3252 | 94 | ||
| XM_015432998.1 | XP_015288484.1 | 5 - 6 | 3452 | 94 | ||
| XM_015432999.1 | XP_015288485.1 | 6 - 7 | 3574 | 94 | ||
| XM_015433000.1 | XP_015288486.1 | 6 - 7 | 3548 | 94 | ||
More Information
Set Membership: |
|

SH3BP4
Genomic Map