Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549832.2 | XP_005549889.1 | 3 - 4 | 839 | 150 | |
| XM_005549833.2 | XP_005549890.1 | 3 - 4 | 961 | 150 | ||
More Information
Set Membership: |
|

KBTBD2
Genomic Map