Assay Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584791.2 | XP_005584848.1 | 1 - 2 | 454 | 78 | |
| XM_015438769.1 | XP_015294255.1 | 1 - 2 | 454 | 78 | ||
More Information
Set Membership: |
|

FAM20A
Genomic Map