Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583828.2 | XP_005583885.1 | 9 - 10 | 4558 | 131 | |
| GenBank mRNA | AB173542.1 | - | 2 - 3 | 547 | 131 | |
More Information
Set Membership: |
|

RNF43
Genomic Map