Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549426.2 | XP_005549483.1 | 7 - 8 | 1142 | 140 | |
| XM_015447229.1 | XP_015302715.1 | 7 - 8 | 1177 | 140 | ||
| XM_015447230.1 | XP_015302716.1 | 6 - 7 | 1472 | 140 | ||
More Information
Set Membership: |
|

TMEM248
Genomic Map