Assay Details
| Gene Symbol: SMG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583863.2 | XP_005583920.1 | 2 - 3 | 1947 | 129 | |
| XM_005583864.2 | XP_005583921.1 | 2 - 3 | 1233 | 129 | ||
More Information
Set Membership: |
|

SMG8
Genomic Map