Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594342.2 | XP_005594399.1 | 3 - 4 | 378 | 89 | |
| XM_005594343.2 | XP_005594400.1 | 3 - 4 | 296 | 89 | ||
| XM_005594344.2 | XP_005594401.1 | 2 - 3 | 213 | 89 | ||
| XM_005594346.2 | XP_005594403.1 | 2 - 3 | 233 | 89 | ||
| GenBank mRNA | AB172396.1 | - | 2 - 3 | 122 | 89 | |
| AB172400.1 | - | 2 - 3 | 122 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005594346.2 transcripts.
Set Membership: |
|

NXT2
Genomic Map