Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566316.2 | XP_005566373.1 | 3 - 4 | 1052 | 71 | |
| GenBank mRNA | AB170805.1 | - | 3 - 4 | 373 | 71 | |
More Information
Set Membership: |
|

TRIM8
Genomic Map