Assay Details
| Gene Symbol: TEX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563005.2 | XP_005563062.1 | 10 - 11 | 9690 | 111 | |
| XM_005563006.2 | XP_005563063.1 | 10 - 11 | 9875 | 111 | ||
| XM_005563007.1 | XP_005563064.1 | 9 - 10 | 9527 | 111 | ||
| XM_015454646.1 | XP_015310132.1 | 11 - 12 | 9660 | 111 | ||
| XM_015454647.1 | XP_015310133.1 | 7 - 8 | 9282 | 111 | ||
More Information
Set Membership: |
|

TEX15
Genomic Map