Assay Details
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566280.2 | XP_005566337.1 | 1 - 2 | 378 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
