Assay Details
| Gene Symbol: PLD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539658.2 | XP_005539715.1 | 9 - 10 | 1562 | 113 | |
| XM_005539659.2 | XP_005539716.1 | 9 - 10 | 1851 | 113 | ||
| XM_005539660.2 | XP_005539717.1 | 9 - 10 | 1499 | 113 | ||
| XM_005539661.2 | XP_005539718.1 | 6 - 7 | 822 | 113 | ||
| XM_015434491.1 | XP_015289977.1 | 9 - 10 | 1857 | 113 | ||
More Information
Set Membership: |
|

PLD5
Genomic Map