Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579574.2 | XP_005579631.1 | 3 - 4 | 869 | 92 | |
| XM_015435686.1 | XP_015291172.1 | 3 - 4 | 1101 | 92 | ||
More Information
Set Membership: |
|

LAYN
Genomic Map