Assay Details
| Gene Symbol: CCDC144NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583126.2 | XP_005583183.1 | 3 - 4 | 828 | 79 | |
| XM_005583128.2 | XP_005583185.1 | 3 - 4 | 828 | 79 | ||
| XM_005583129.2 | XP_005583186.1 | 3 - 4 | 828 | 79 | ||
| XM_005583130.2 | XP_005583187.1 | 3 - 4 | 828 | 79 | ||
| GenBank mRNA | AB074451.1 | - | 3 - 4 | 875 | 79 | |
More Information
Set Membership: |
|

CCDC144NL
Genomic Map