Assay Details
| Gene Symbol: MDGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561185.2 | XP_005561242.1 | 4 - 5 | 954 | 101 | |
| XM_015453753.1 | XP_015309239.1 | 4 - 5 | 2729 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015453753.1 transcripts.
Set Membership: |
|

MDGA2
Genomic Map