Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591592.2 | XP_005591649.1 | 4 - 5 | 754 | 62 | |
| XM_005591593.2 | XP_005591650.1 | 4 - 5 | 738 | 62 | ||
| XM_005591594.2 | XP_005591651.1 | 4 - 5 | 784 | 62 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map