Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549976.2 | XP_005550033.1 | 2 - 3 | 345 | 99 | |
| XM_015447634.1 | XP_015303120.1 | 2 - 3 | 345 | 99 | ||
| XM_015447635.1 | XP_015303121.1 | 2 - 3 | 345 | 99 | ||
| XM_015447636.1 | XP_015303122.1 | 2 - 3 | 345 | 99 | ||
More Information
Set Membership: |
|

FAM221A
Genomic Map