Assay Details
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584185.1 | XP_005584242.1 | 7 - 8 | 1876 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584185.1 | XP_005584242.1 | 7 - 8 | 1876 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |