Assay Details
| Gene Symbol: KCNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578336.2 | XP_005578393.1 | 1 - 2 | 555 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005578336.2 transcripts.
Set Membership: |
|

KCNA4
Genomic Map