Assay Details
Gene Symbol: PRTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587337.2 | XP_005587394.1 | 2 | 259 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
