Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565238.2 | XP_005565295.1 | 7 | 4026 | 159 | |
| XM_005565239.2 | XP_005565296.1 | 7 | 3366 | 159 | ||
More Information
Set Membership: |
|

MMRN2
Genomic Map