Assay Details
| Gene Symbol: ACOT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561722.2 | XP_005561779.1 | 1 | 395 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005561722.2 transcripts.
Set Membership: |
|

ACOT4
Genomic Map