Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441100.1 | XP_015296586.1 | 9 | 2965 | 96 | |
| XM_015441101.1 | XP_015296587.1 | 8 | 2882 | 96 | ||
| XM_015441102.1 | XP_015296588.1 | 9 | 2915 | 96 | ||
| XM_015441103.1 | XP_015296589.1 | 7 | 2554 | 96 | ||
| XM_015441104.1 | XP_015296590.1 | 7 | 2733 | 96 | ||
More Information
Set Membership: |
|

ZNF45
Genomic Map