Assay Details
| Gene Symbol: IRF2BPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561872.2 | XP_005561929.1 | 2 | 3188 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRF2BPL| Gene Symbol: IRF2BPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561872.2 | XP_005561929.1 | 2 | 3188 | 113 | |
Set Membership: |
|