Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586777.2 | XP_005586834.1 | 5 - 6 | 1407 | 64 | |
| XM_005586778.2 | XP_005586835.1 | 4 - 5 | 1299 | 64 | ||
| XM_005586779.2 | XP_005586836.1 | 3 - 4 | 1206 | 64 | ||
| XM_015439921.1 | XP_015295407.1 | 4 - 5 | 2321 | 64 | ||
More Information
Set Membership: |
|

ST8SIA5
Genomic Map