Assay Details
| Gene Symbol: SPATA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569193.2 | XP_005569250.1 | 1 - 2 | 550 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA25| Gene Symbol: SPATA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569193.2 | XP_005569250.1 | 1 - 2 | 550 | 84 | |
Set Membership: |
|