Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283407.1 | NP_001270336.1 | 4 - 5 | 486 | 59 | |
| GenBank mRNA | AB179206.1 | - | 4 - 5 | 486 | 59 | |
More Information
Set Membership: |
|

RRP7A
Genomic Map